Learn More
Instech Tether for magnetic 1 channel rat Vascular Access Button (white port), 12in, tube fits 22ga
SDP
Catalog No. NC2605414
Encompass
One channel magnetic Vascular Access Button tether kit for rats. Mates with one channel Vascular Access Button for rats.
Includes:
  • One channel rat tether (VABR1T/22), plastic swivel (375/22PS)
  • 24” of CoEx tubing
  • One luer stub (LS22)
Catalog No. NC2605414 Supplier Instech Supplier No. VABR1T/22
One channel magnetic Vascular Access Button tether kit for rats. Mates with one channel Vascular Access Button for rats.
Includes:
  • One channel rat tether (VABR1T/22), plastic swivel (375/22PS)
  • 24” of CoEx tubing
  • One luer stub (LS22)