Learn More
Mopec Tether Stakes
SDP
Catalog No. 22110491 Shop All Mopec Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
22-110-491 4/Pk.
1 options
Catalog No. 22-110-491 Supplier Mopec Supplier No. FS890
Add to Cart
Add to Cart
Ground stakes designed to securely anchor barrier systems and help prevent movement, shifting, or tipping during use.
  • Options and Accessories
  • FS871: 1000 Barrier System
  • FS873: Quad Barrier System
  • FS874: Octo Barrier System
  • FS785: Copper Barrier System
Specifications
Product Type Tether Stakes
Quantity 4/Pk.