Learn More
GENEPATH DIAGNOSTICS INC TetM quantitative standards for assay calibration and validation. Provided at high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL) concentrations (RUO)
Catalog No. NC3949385
Encompass_Preferred
TetM quantitative standards are designed to support precise assay calibration and validation in molecular diagnostics. Supplied at three concentrations—high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL)—these standards enable accurate assessment of assay performance across a dynamic range. They are ideal for sensitivity testing, method development, quality control, and proficiency testing, ensuring reproducible and reliable detection of TetM target sequences. Ready-to-use and stable, these standards streamline workflow integration while maintaining high confidence in molecular assay results. Suitable for research, clinical, and surveillance applications, they provide a trusted reference for laboratories aiming for accuracy and consistency in nucleic acid detection. (RUO)
Catalog No. NC3949385 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10830120050
TetM quantitative standards are designed to support precise assay calibration and validation in molecular diagnostics. Supplied at three concentrations—high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL)—these standards enable accurate assessment of assay performance across a dynamic range. They are ideal for sensitivity testing, method development, quality control, and proficiency testing, ensuring reproducible and reliable detection of TetM target sequences. Ready-to-use and stable, these standards streamline workflow integration while maintaining high confidence in molecular assay results. Suitable for research, clinical, and surveillance applications, they provide a trusted reference for laboratories aiming for accuracy and consistency in nucleic acid detection. (RUO)