Learn More
Cayman Chemical Tetnus ToxIn CFragment10ug
SDP
Catalog No. 502993837
Encompass_Preferred
A C-terminal fragment of a neurotoxin; binds to gangliosides on presynaptic terminals at the neuromuscular junction
Catalog No. 50-299-3837 Supplier Cayman Chemical Supplier No. 2322210 UG
A C-terminal fragment of a neurotoxin; binds to gangliosides on presynaptic terminals at the neuromuscular junction