Learn More
BIOHIPPO TetOn h/m MYC-sh-BSD
SDP
Catalog No. 5000331459
Encompass
TetOn h/m MYC-sh-BSD shRNA lentiviral vector expressing with Blasticidin selection for studies in Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m c-MYC shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse MYC proto-oncogene bHLH transcription factor (MYC) also known as c-MYC or cMYC The shRNA has been validated to meet or exceed 70% c-MYC knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31459 Supplier BIOHIPPO Supplier No. BHV194001311
TetOn h/m MYC-sh-BSD shRNA lentiviral vector expressing with Blasticidin selection for studies in Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m c-MYC shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse MYC proto-oncogene bHLH transcription factor (MYC) also known as c-MYC or cMYC The shRNA has been validated to meet or exceed 70% c-MYC knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs