Learn More
ATCC tet-on spiking HEK cells; Embryonic Kidney; Human (Homo sapiens)
Catalog No. 502382554
Encompass
tet-on spiking HEK cells is a cell line exhibiting epithelial and/or rounded morphology. It was derived from an existing cell line that was originally isolated from embryonic kidney tissue. It has applications for synthetic electrophysiological systems and is useful in testing channel rhodopsins and voltage indicators.
Catalog No. 50-238-2554 Supplier ATCC Supplier No. CRL3479
tet-on spiking HEK cells is a cell line exhibiting epithelial and/or rounded morphology. It was derived from an existing cell line that was originally isolated from embryonic kidney tissue. It has applications for synthetic electrophysiological systems and is useful in testing channel rhodopsins and voltage indicators.