Learn More
Apexbio Technology LLC Tetraethylenepentamine 5HCl, 50mg. Cas: 4961-41-5 MFCD: MFCD00083503. polyamine catalyst for the bisulfite reaction of cytosine
SDP
Supplier:  Apexbio Technology LLC A853550
Encompass_Preferred
Tetraethylenepentamine 5HCl Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5158