Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tetra Core Set and NuPAGE™ Bis-Tris 4-12% Gels Combos

Catalog No. NP032TS12W
Encompass_Preferred
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
NP032TS12W 12-well
NP032TS10W 10-well
NP032TS15W 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NP032TS12W Supplier Invitrogen™ Supplier No. NP032TS12W
Save 22% on Invitrogen Gels, Reagents, and More
Use promo code GELS22
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Tetra Electrophoresis Core Set and NuPAGE Bis-Tris 4-12% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of NuPAGE Bis-Tris gels. Easily swap and run up to four NuPAGE gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank, achieving superior protein resolution and reproducibility.

The Invitrogen Tetra Electrophoresis Core Set and NuPAGE Bis-Tris 4-12% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of NuPAGE Bis-Tris gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of NuPAGE Bis-Tris 4-12% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra Electrophoresis cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both the primary and companion cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About NuPAGE Bis-Tris gels

Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use. Each box contains 10 gels.

TRUSTED_SUSTAINABILITY

Specifications

Adapters Tetra Electrophoresis Cores
Content And Storage Store at 4°C to 25°C. Do not freeze.
Buffer NuPAGE MES SDS Running Buffer, NuPAGE MOPS SDS Running Buffer
Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
Sample Loading Volume Up to 20 μL
Separation of Protein
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 12-well
Mode of Separation Molecular Weight
Quantity Tetra Core Set, 3 boxes of gels (30 gels)
Shipping Condition Room Temperature
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line NuPAGE
Product Type Electrophoresis Cores and Precast Protein Gels
For Use With (Equipment) Bio-Rad Mini Protean Tanks
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.