Learn More
Medchemexpress LLC Tetra-N-acetylchitotetraose | 2706-65-2 | 1 MG
SDP
Catalog No. 5000417700
Encompass_Preferred
Tetra-N-acetylchitotetraose elicits plant defense systems and is a component of hpo-chitoo gosacchaπdes (LCOs) secreted from Rhizobia. It also serves as a substrate for the Rhizobium leguminosarum nodulation protein NodB, a CO deacetylase.
  • Elicits plant defense systems
  • Component of hpo-chitoo gosacchaπdes (LCOs) secreted from Rhizobia
  • Substrate for the Rhizobium leguminosarum nodulation protein NodB, a CO deacetylase
Catalog No. 50-004-17700 Supplier Medchemexpress LLC Supplier No. HYN76981MG
Tetra-N-acetylchitotetraose elicits plant defense systems and is a component of hpo-chitoo gosacchaπdes (LCOs) secreted from Rhizobia. It also serves as a substrate for the Rhizobium leguminosarum nodulation protein NodB, a CO deacetylase.
  • Elicits plant defense systems
  • Component of hpo-chitoo gosacchaπdes (LCOs) secreted from Rhizobia
  • Substrate for the Rhizobium leguminosarum nodulation protein NodB, a CO deacetylase