Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium borohydride, 97%

Catalog No. AAA1749430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1749430 250 g
AAA1749409 10 g
AAA1749418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1749430 Supplier Thermo Scientific Chemicals Supplier No. A1749430
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33725-74-5 | C16H40BN | 257.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.31
MDL Number MFCD00012035
InChI Key GMBOFJFPOCGSOI-UHFFFAOYSA-N
Synonym tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
PubChem CID 9881569
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 125°C to 131°C
Flash Point 140°C (284°F)
Linear Formula [CH3(CH2)3]4NBH4
Quantity 250 g
UN Number UN3131
Sensitivity Moisture sensitive
IUPAC Name boron(1-);tetrabutylazanium
Formula Weight 257.31
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium borohydride
Hazard Category H261-H314
Hazard Statement GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
EINECSNumber 251-658-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.