Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium fluoride, 1M soln. in THF

Catalog No. p-7080862
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10588AC 25 mL
AAA10588AE 100 mL
AAA10588AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10588AC Supplier Thermo Scientific Chemicals Supplier No. A10588AC
Only null left
Add to Cart

CAS: 429-41-4 | C16H36FN | 261.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride, 1M soln. in THF is used as a reactant for the preparation of triple monoamine reuptake inhibitors as a new generation of antidepressants, oligoribonucleotides with phosphonate-modified linkages, conjugated dienoic acid esters using Suzuki coupling reactions and polyacetylenic glucosides. It plays an important role in aryl alkyl alcohols through Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride. It is also used as a catalyst for the silylation reactions. In addition, it is used as a source of fluoride ion in organic solvents as well as for the deprotection of silyl ether groups.

Solubility
Miscible with terahydrofuran, acetonitrile, dimethyl sulfoxide and organic solvents.

Notes
Store in a cool place. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
IUPAC Name tetrabutylazanium;fluoride
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.903
Flash Point −17°C (1°F)
Linear Formula [CH3(CH2)3]4NF
Quantity 25 mL
UN Number UN2924
Beilstein 3570522
Sensitivity Hygroscopic
Solubility Information Miscible with terahydrofuran,acetonitrile,dimethyl sulfoxide and organic solvents.
Formula Weight 261.47
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Tetra-n-butylammonium fluoride
Show More Show Less
Hazard Category H225-H314-H335-H336-H351-H373
Hazard Statement GHS H Statement
H225-H314-H351-H302-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 207-057-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.