Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium hexafluorophosphate, 98%

Catalog No. p-4258672
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC352580250 25 g Glass bottle
AC352581000 100 g Glass bottle
AC352585000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC352580250 Supplier Thermo Scientific Chemicals Supplier No. 352580250
Only null left
Add to Cart

CAS: 3109-63-5 | C16H36F6NP | 387.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3109-63-5
Molecular Formula C16H36F6NP
Molecular Weight (g/mol) 387.44
MDL Number MFCD00011748
InChI Key BKBKEFQIOUYLBC-UHFFFAOYSA-N
Synonym tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate
PubChem CID 165075
IUPAC Name tetrabutylazanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 244.0°C to 246.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 25 g
Assay Percent Range 98%
Linear Formula [CH3(CH2)3]4N(PF6)
Formula Weight 387.44
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Tetra-n-butylammonium hexafluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 221-472-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.