Learn More
Crescent Chemical Co Inc TETRA-N-BUTYLAMMONIUM-HYDROGEN
SDP
Catalog No. 50248180
Encompass_Preferred
Tetra-n-butylammonium-hydrogensulfate analytical grade For ion-pair reversed phase chromatography of ribonucleotides. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35854.01 CAS 32503-27-8
Catalog No. 50-248-180 Supplier Crescent Chemical Co Inc Supplier No. SE35854.01
Tetra-n-butylammonium-hydrogensulfate analytical grade For ion-pair reversed phase chromatography of ribonucleotides. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35854.01 CAS 32503-27-8