Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium hydroxide, Reagent Grade, 31% in methanol, Thermo Scientific™

Catalog No. AA1626306
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA1626306 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1626306 Supplier Thermo Scientific Chemicals Supplier No. 1626306

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

As a basic nonaqueous titrant

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2052-49-5
Density 0.830g/mL
Flash Point 13.9° (57°F)
Odor Alcohol-like
Molecular Formula C16H37NO
Refractive Index 1.3775
Quantity 5g
MDL Number MFCD00009425
UN Number UN2924
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.48
Grade Reagent
Concentration or Composition (by Analyte or Components) 31% in methanol
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H224-H300-H310-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.