Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetra-n-butylammonium tetrafluoroborate, 99%

Click to view available options
Quantity:
5g
25g
429-42-5
C16H36BF4N
329.275
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate,tetra-n-butylammonium tetrafluoroborate,tetrabutylammonium fluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-,tetrabutyl ammonium tetrafluoroborate,tetrabutylammonium ion tetrafluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1,tbabf4,acmc-209jry,tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
[B-](F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
429-42-5
C16H36BF4N
329.275
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate,tetra-n-butylammonium tetrafluoroborate,tetrabutylammonium fluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-,tetrabutyl ammonium tetrafluoroborate,tetrabutylammonium ion tetrafluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1,tbabf4,acmc-209jry,tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
[B-](F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Applications
As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

Solubility
Slightly soluble in Water, methanol( 0.1 g/mL), ethanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 25g
Odor Odorless
Melting Point 159°C to 162°C
Linear Formula [CH3(CH2)3]4NBF4
Beilstein 3577508
Solubility Information Slightly soluble in Water,methanol( 0.1g/mL),ethanol (100mg/ml).
Formula Weight 329.27
Percent Purity 99%
Chemical Name or Material Tetra-n-butylammonium tetrafluoroborate

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-058-8

TSCA : Yes

Recommended Storage : Ambient temperatures