Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium tetrafluoroborate, Thermo Scientific™

Catalog No. AA1757904
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1757904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1757904 Supplier Thermo Scientific Chemicals Supplier No. 1757904

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-42-5
Molecular Formula C16H36BF4N
Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
PubChem CID 67932
IUPAC Name tetrabutylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 157°C to 160°C
Odor Odorless
Linear Formula [CH3(CH2)3]4NBF4
Quantity 2 g
Beilstein 3577508
Formula Weight 329.27
Physical Form Powder
Chemical Name or Material Tetra-n-butylammonium tetrafluoroborate
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.