Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butyltin, 96%

Catalog No. AC137980050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137980050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC137980050 Supplier Thermo Scientific Chemicals Supplier No. 137980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1461-25-2 | C16H36Sn | 347.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-25-2
MDL Number MFCD00009422
InChI Key AFCAKJKUYFLYFK-UHFFFAOYSA-N
Synonym tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
PubChem CID 15098
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Melting Point -97°C
Color Colorless to Straw
Density 1.0570g/mL
Boiling Point 145°C (10.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C16H36Sn
Refractive Index 1.4725 to 1.4745
Linear Formula [CH3(CH2)3]4Sn
Quantity 5 g
Specific Gravity 1.057
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 347.14
Viscosity 4 mPa.s (20°C)
Formula Weight 347.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-960-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.