Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-heptylammonium iodide, 99%, Thermo Scientific™

Catalog No. AAA1664222
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1664222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1664222 Supplier Thermo Scientific Chemicals Supplier No. A1664222

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3535-83-9
Molecular Formula C28H60IN
Molecular Weight (g/mol) 537.699
MDL Number MFCD00041984
InChI Key KCSOHLKZTZMKQA-UHFFFAOYSA-M
Synonym tetraheptylammonium iodide, tetra-n-heptylammonium iodide, 1-heptanaminium, n,n,n-triheptyl-, iodide, tetraheptylazanium iodide, 1-heptanaminium, n,n,n-triheptyl-, iodide 1:1, ammonium, tetraheptyl-, iodide, acmc-1aeuw, 1-heptanaminium,n,n-triheptyl-, iodide, tetraheptylammonium iodide at
PubChem CID 77071
IUPAC Name tetraheptylazanium;iodide
SMILES CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[I-]

Specifications

Melting Point 122°C to 124°C
Quantity 100 g
Beilstein 4167115
Sensitivity Hygroscopic; Light sensitive
Formula Weight 537.7
Percent Purity 99%
Chemical Name or Material Tetra-n-heptylammonium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-570-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.