Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-octylammonium Iodide 98.0+%, TCI America™
SDP

Catalog No. T11555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11555G Supplier TCI America Supplier No. T11555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16829-91-7
Molecular Formula C32H68IN
Molecular Weight (g/mol) 593.807
MDL Number MFCD00059978
InChI Key KGPZZJZTFHCXNK-UHFFFAOYSA-M
PubChem CID 11180877
IUPAC Name tetraoctylazanium;iodide
SMILES CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[I-]

Specifications

Melting Point 129°C
Color White
Quantity 5 g
Formula Weight 593.81
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetra-n-octylammonium Iodide
EINECSNumber (9)-1971
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.