Learn More
Medchemexpress LLC Tetra-n-pentylammonium iodide | 2498-20-6 | MFCD00041980 | >98.0% | 425.47 g/mol | C20H44IN | 25 G
SDP
Catalog No. 5000283095
Encompass_Preferred
Tetraamylammonium iodide is a quaternary ammonium salt (also known as tetrapentylammonium iodide) supplied as a white solid for laboratory research. It is commonly used as a phase-transfer catalyst, ion-pairing agent, or non-coordinating counterion in organic synthesis and biochemical workflows. Typical reported specifications include purity >98.0%, molecular formula C20H44IN, and molecular weight 425.47 g/mol.
  • High purity suitable for research applications.
  • White, solid salt form for ease of handling and weighing.
  • Effective as a phase-transfer catalyst in organic reactions.
  • Useful as an ion-pairing agent and non-coordinating counterion.
  • Compatible with common organic solvents and standard laboratory procedures.
  • Packaged in small-scale quantities suitable for bench-top use.
Catalog No. 50-002-83095 Supplier Medchemexpress LLC Supplier No. HYW12765925G
Tetraamylammonium iodide is a quaternary ammonium salt (also known as tetrapentylammonium iodide) supplied as a white solid for laboratory research. It is commonly used as a phase-transfer catalyst, ion-pairing agent, or non-coordinating counterion in organic synthesis and biochemical workflows. Typical reported specifications include purity >98.0%, molecular formula C20H44IN, and molecular weight 425.47 g/mol.
  • High purity suitable for research applications.
  • White, solid salt form for ease of handling and weighing.
  • Effective as a phase-transfer catalyst in organic reactions.
  • Useful as an ion-pairing agent and non-coordinating counterion.
  • Compatible with common organic solvents and standard laboratory procedures.
  • Packaged in small-scale quantities suitable for bench-top use.