Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide, ACS, 99.5% min

Catalog No. AA7113709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7113709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA7113709 Supplier Thermo Scientific Chemicals Supplier No. 07113709
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2176-98-9 | C12H28Sn | 291.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2176-98-9
Molecular Formula C12H28Sn
Molecular Weight (g/mol) 291.07
MDL Number MFCD00015196
InChI Key OIQCWAIEHVRCCG-UHFFFAOYSA-N
Synonym tetra-n-propyltin, tetrapropyltin, stannane, tetrapropyl, tin, tetrapropyl, tin tetra-n-propyl, tetra n-propyltin, tetrapropyl tin, c3h7 4sn, oiqcwaiehvrccg-uhfffaoysa
PubChem CID 16585
IUPAC Name tetrapropylstannane
SMILES CCC[Sn](CCC)(CCC)CCC

Specifications

Color Colorless
Density 1.11 g/mL
Boiling Point 222°C
Odor Unpleasant
Refractive Index 1.4745
Linear Formula (CH3CH2CH2)4Sn
Quantity 10 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 291.04
Physical Form Liquid
Chemical Name or Material Tetra-n-propyltin
Show More Show Less
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 218-536-0
RTECSNumber WH8636500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.