Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-O-acetyl-beta-D-ribopyranose 98.0+%, TCI America™
SDP

Catalog No. R00651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00651G Supplier TCI America Supplier No. R00651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4049-34-7
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00006596
InChI Key MJOQJPYNENPSSS-LPWJVIDDSA-N
Synonym tetra-o-acetyl-beta-d-ribopyranose, 2s,3r,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4-tetra-o-acetyl-b-d-ribopyranose, beta-d-ribopyranose 1,2,3,4-tetraacetate, 3r,4r,5r,6s-4,5,6-triacetyloxyoxan-3-yl acetate, 2s,3r,4r,5r-2,4,5-tris acetyloxy oxan-3-yl acetate, beta-d-ribopyranose tetraacetate, mjoqjpynenpsss-lpwjviddsa, 1,2,3,4-tetra-o-acetyl-, a-d-ribopyranose, b-d-ribopyranose 1,2,3,4-tetraacetate
PubChem CID 97336
IUPAC Name [(3R,4R,5R,6S)-4,5,6-triacetyloxyoxan-3-yl] acetate
SMILES CC(=O)OC1COC(C(C1OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 318.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetra-O-acetyl-beta-D-ribopyranose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.