Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetra(p-bromophenyl)porphyrin | 29162-73-0 | 95.0% | 930.32 | C44H26Br4N4 | 5 G
SDP

Catalog No. 5000300079
Encompass_Preferred

Tetra(p-bromophenyl)porphyrin is a brominated tetrakisphenylporphyrin used as a fluorescent dye and as a synthetic intermediate in porphyrin chemistry. It is employed as a porphyrin scaffold for derivatization and photophysical studies.

  • Fluorescent tetrakisphenylporphyrin with four p-bromophenyl groups.
  • CAS number 29162-73-0 and formula C44H26Br4N4.
  • Molecular weight 930.32 g·mol-1.
  • Light blue to blue solid, purity ~95.0%.
  • Suitable as a building block for perfluoroalkyl-substituted porphyrins and other derivatives.
  • Store protected from light and under inert atmosphere; cold storage recommended for solutions.

Catalog No. 50-003-00079 Supplier Medchemexpress LLC Supplier No. HYW0351385G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetra(p-bromophenyl)porphyrin is a brominated tetrakisphenylporphyrin used as a fluorescent dye and as a synthetic intermediate in porphyrin chemistry. It is employed as a porphyrin scaffold for derivatization and photophysical studies.

  • Fluorescent tetrakisphenylporphyrin with four p-bromophenyl groups.
  • CAS number 29162-73-0 and formula C44H26Br4N4.
  • Molecular weight 930.32 g·mol-1.
  • Light blue to blue solid, purity ~95.0%.
  • Suitable as a building block for perfluoroalkyl-substituted porphyrins and other derivatives.
  • Store protected from light and under inert atmosphere; cold storage recommended for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.