Learn More
Strem, An Ascensus Company CAS# 20634-12-2. 5g. Tetraammineplatinum(II) nitrate, 99%. MFCD00011624
SDP
Catalog No. 5090113330
Encompass
CAS# 20634-12-2. 5g. Tetraammineplatinum(II) nitrate, 99%. MFCD00011624. Molecular Weight: 387.22. Molecular Formula: [Pt(NH3)4](NO3)2. Color/form: white to off-white pwdr. Strem# 78-2010. http://www.strem.com/catalog/v/78-2010/
Catalog No. 50-901-13330 Supplier Strem, An Ascensus Company Supplier No. 7820105G
CAS# 20634-12-2. 5g. Tetraammineplatinum(II) nitrate, 99%. MFCD00011624. Molecular Weight: 387.22. Molecular Formula: [Pt(NH3)4](NO3)2. Color/form: white to off-white pwdr. Strem# 78-2010. http://www.strem.com/catalog/v/78-2010/