Learn More
Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min
Description
Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 262°C (decomposition) |
| Color | White to Yellow |
| Physical Form | Crystals |
| Quantity | 0.25 g |
| Assay Percent Range | 99.99% (metals basis) |
| Solubility Information | Soluble in water. |
| IUPAC Name | platinum(2+) tetraamine dinitrate |
| Formula Weight | 387.21 |
| Chemical Name or Material | Tetraammineplatinum(II) nitrate |
Safety and Handling
| missing translation for 'healthHazard1' | H272-H315-H317-H319-H335 |
| missing translation for 'healthHazard3' | P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1; Packaging Group: II |
| EINECSNumber | 243-929-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.