Learn More
Medchemexpress LLC Tetraamylammonium chloride | 4965-17-7 | MFCD00011857 | 99.9% | 334.02 g·mol⁻¹ | C20H44ClN | 250 MG
SDP
Catalog No. 5000283529
Encompass_Preferred
Tetraamylammonium chloride is a quaternary ammonium salt used primarily as a phase transfer catalyst and as an intermediate in organic synthesis. It is supplied as a high-purity solid suitable for research and synthetic applications where transfer of ionic species between immiscible phases is required.
  • High purity (99.9%).
  • Solid form for ease of handling and storage.
  • Used as a phase transfer catalyst in organic synthesis.
  • Serves as an intermediate for synthesis of quaternary ammonium compounds.
  • Storage: keep dry and cool; in solvent, store at -80°C up to 2 years or -20°C up to 1 year.
Catalog No. 50-002-83529 Supplier Medchemexpress LLC Supplier No. HYW127637250MG
Tetraamylammonium chloride is a quaternary ammonium salt used primarily as a phase transfer catalyst and as an intermediate in organic synthesis. It is supplied as a high-purity solid suitable for research and synthetic applications where transfer of ionic species between immiscible phases is required.
  • High purity (99.9%).
  • Solid form for ease of handling and storage.
  • Used as a phase transfer catalyst in organic synthesis.
  • Serves as an intermediate for synthesis of quaternary ammonium compounds.
  • Storage: keep dry and cool; in solvent, store at -80°C up to 2 years or -20°C up to 1 year.