Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraamylammonium Chloride 98.0+%, TCI America™
SDP

Catalog No. T14335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14335G 5 g
T143325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14335G Supplier TCI America Supplier No. T14335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4965-17-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.029
MDL Number MFCD00011857
InChI Key SXAWRMKQZKPHNJ-UHFFFAOYSA-M
Synonym Tetrapentylammonium Chloride
PubChem CID 78667
IUPAC Name tetrapentylazanium;chloride
SMILES CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-]

Specifications

Color White
Quantity 5 g
UN Number 2811
Formula Weight 334.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraamylammonium Chloride
EINECSNumber (2)-0186
RTECSNumber RZ9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.