Learn More
Bachem H-Arg-Arg-Arg-Arg-OH
SDP
Supplier:  Bachem 4042835.0025
Encompass_Preferred
H-Arg-Arg-Arg-Arg-OH Acetate, 26791-46-8, C24H50N16O5, Mr 642.77, 25mg. Tetraarginine acetate salt. Tetraarginine, RRRR, is one of the commonly observed repeat units in protamine molecules.
Catalog No. 50-194-7513