Learn More
Cayman Chemical Tetrabenazine, Purity: ≥98% (mixture of enantiomers), CAS: 58-46-8, 100 mg
SDP
Catalog No. 502983847
Encompass_Preferred
An inhibitor of VMAT2 that is selective over VMAT1 (Kis = 97 and >20,000 nM, respectively, in a serotonin uptake assay); dose-dependently reduces levels of the monoamines norepinephrine, dopamine, and 5-HT in rat brain; has been used to induce depressive-like behavior in animal models; improves performance in balance beam and rotarod tests and prevents decreases in the number of striatal MSNs in a YAC128 transgenic mouse model of Huntington’s disease at 5 mg/kg
Catalog No. 50-298-3847 Supplier Cayman Chemical Supplier No. 20380100 MG
An inhibitor of VMAT2 that is selective over VMAT1 (Kis = 97 and >20,000 nM, respectively, in a serotonin uptake assay); dose-dependently reduces levels of the monoamines norepinephrine, dopamine, and 5-HT in rat brain; has been used to induce depressive-like behavior in animal models; improves performance in balance beam and rotarod tests and prevents decreases in the number of striatal MSNs in a YAC128 transgenic mouse model of Huntington’s disease at 5 mg/kg