Learn More
Cayman Chemical TetrabenazIn-d6 10mg
SDP
Catalog No. 502936767
Encompass_Preferred
An internal standard for the quantification of tetrabenazine by GC- or LC-MS
Catalog No. 50-293-6767 Supplier Cayman Chemical Supplier No. 2681110 MG
An internal standard for the quantification of tetrabenazine by GC- or LC-MS