Learn More
Selleck Chemical LLC Tetrabenazine (Xenazine) S1789-10mg
SDP
Catalog No. 503110871
Encompass_Preferred
Tetrabenazine(Xenazine Ro 1-9569) acts primarily as a reversible high-affinity inhibitor of mono-amine uptake into granular vesicles of presynaptic neurons by binding selectively to VMAT-2 Also blocks dopamine D2 receptors but this affinity is 1 000-fold lower than its affinity for VMAT-2
Catalog No. 50-311-0871 Supplier Selleck Chemical LLC Supplier No. S178910MG
Tetrabenazine(Xenazine Ro 1-9569) acts primarily as a reversible high-affinity inhibitor of mono-amine uptake into granular vesicles of presynaptic neurons by binding selectively to VMAT-2 Also blocks dopamine D2 receptors but this affinity is 1 000-fold lower than its affinity for VMAT-2