Learn More
Medchemexpress LLC Tetrabenazine-d6 | 1392826-25-3 | 323.46 | C19H21D6NO3 | 10 MG
SDP
Catalog No. 5000298295
Encompass_Preferred
Tetrabenazine-d6 is a deuterium-labeled analogue of tetrabenazine provided as a research reference standard for analytical and preclinical applications. It serves as a tracer or internal standard for quantitative analyses (LC-MS, GC-MS, NMR), and is used in studies of vesicular monoamine transporter activity and hyperkinetic movement disorder research.
  • Deuterium-labeled isotopologue for analytical internal standard use.
  • Useful for LC-MS, GC-MS, and NMR quantification.
  • Targeted for monoamine transporter (VMAT) research.
  • Soluble in DMSO; limited solubility in water (may require ultrasonic).
  • Supplied as a small-quantity research reference (10 mg).
Catalog No. 50-002-98295 Supplier Medchemexpress LLC Supplier No. HYB0590S10MG
Tetrabenazine-d6 is a deuterium-labeled analogue of tetrabenazine provided as a research reference standard for analytical and preclinical applications. It serves as a tracer or internal standard for quantitative analyses (LC-MS, GC-MS, NMR), and is used in studies of vesicular monoamine transporter activity and hyperkinetic movement disorder research.
  • Deuterium-labeled isotopologue for analytical internal standard use.
  • Useful for LC-MS, GC-MS, and NMR quantification.
  • Targeted for monoamine transporter (VMAT) research.
  • Soluble in DMSO; limited solubility in water (may require ultrasonic).
  • Supplied as a small-quantity research reference (10 mg).