Learn More
Medchemexpress LLC Tetrabenazine-d6 | 1392826-25-3 | 98.2% | 100 MG
SDP
Catalog No. 5000408549
Encompass_Preferred
Tetrabenazine-d6, also known as Deutetrabenazine, is a deuterium-labeled compound. It is approved worldwide for the research of Huntington's disease and other hyperkinetic movement disorders.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Extends the half-life in vivo
  • Improves oral bioavailability
  • Improves metabolic characteristics
  • Improves drug safety
  • Preserves therapeutic properties
Catalog No. 50-004-08549 Supplier Medchemexpress LLC Supplier No. HYB0590S100MG
Tetrabenazine-d6, also known as Deutetrabenazine, is a deuterium-labeled compound. It is approved worldwide for the research of Huntington's disease and other hyperkinetic movement disorders.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Extends the half-life in vivo
  • Improves oral bioavailability
  • Improves metabolic characteristics
  • Improves drug safety
  • Preserves therapeutic properties