Learn More
Medchemexpress LLC Tetrabenazine-d6 (Ro 1-9569-d6) | 1392826-25-3 | ≥99.0% | 50 MG
SDP
Catalog No. 5000408332
Encompass_Preferred
Tetrabenazine-d6 (Deutetrabenazine) is a deuterium-labeled Tetrabenazine (HY-B0590). It is the first deuterium approved worldwide for the research of Huntington's disease, or other hyperkinetic movement disorders. This compound is for research use only and not sold to patients. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterated compounds are expected to retain similar biochemical potency and selectivity to hydrogen analogs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Extends half-life in vivo
  • Improves oral bioavailability
  • Improves metabolic characteristics
  • Improves drug safety
  • Preserves therapeutic properties
Catalog No. 50-004-08332 Supplier Medchemexpress LLC Supplier No. HYB0590S50MG
Tetrabenazine-d6 (Deutetrabenazine) is a deuterium-labeled Tetrabenazine (HY-B0590). It is the first deuterium approved worldwide for the research of Huntington's disease, or other hyperkinetic movement disorders. This compound is for research use only and not sold to patients. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterated compounds are expected to retain similar biochemical potency and selectivity to hydrogen analogs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Extends half-life in vivo
  • Improves oral bioavailability
  • Improves metabolic characteristics
  • Improves drug safety
  • Preserves therapeutic properties