Learn More
TARGETMOL CHEMICALS INC TETRABENAZINE METABOLITE 5MG
SDP
Catalog No. 502581378
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tetrabenazine Metabolite ((-)-(beta)-HTBZ) a vesicles monoamine transporter 2 (VMAT2) inhibitor is an active metabolite of Tetrabenazine. Tetrabenazine Metabolite has a high affinity for VMAT2 with a Ki of 13.4 nM. Tetrabenazine Metabolite is often used to study chorea associated with Huntington's disease and other hyperactivity disorders. purity: 95%
Catalog No. 50-258-1378 Supplier TARGETMOL CHEMICALS INC Supplier No. T131335MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tetrabenazine Metabolite ((-)-(beta)-HTBZ) a vesicles monoamine transporter 2 (VMAT2) inhibitor is an active metabolite of Tetrabenazine. Tetrabenazine Metabolite has a high affinity for VMAT2 with a Ki of 13.4 nM. Tetrabenazine Metabolite is often used to study chorea associated with Huntington's disease and other hyperactivity disorders. purity: 95%