Learn More
Abcam Tetrabenazine, VMAT2 inhibitor, 25MG
Catalog No. 502704902
Encompass_Preferred
MW 317.42 g/mol. Potent, reversible VMAT2 inhibitor (IC₅₀ = 3.2 nM, Ki = 100 nM). Depletes monoamines stores and decreases uptake into synaptic vesicles. Shows antihyperkinetic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4902 Supplier Abcam Supplier No. AB14616625MG
Add to Cart
Add to Cart
MW 317.42 g/mol. Potent, reversible VMAT2 inhibitor (IC₅₀ = 3.2 nM, Ki = 100 nM). Depletes monoamines stores and decreases uptake into synaptic vesicles. Shows antihyperkinetic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement