Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromobisphenol A 98.0+%, TCI America™
SDP

Catalog No. T0032500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0032500G 500 g
T003225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0032500G Supplier TCI America Supplier No. T0032500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79-94-7
Molecular Formula C15H12Br4O2
Molecular Weight (g/mol) 543.875
MDL Number MFCD00013962
InChI Key VEORPZCZECFIRK-UHFFFAOYSA-N
Synonym tetrabromobisphenol a, 3,3',5,5'-tetrabromobisphenol a, bromdian, 4,4'-propane-2,2-diyl bis 2,6-dibromophenol, 4,4'-isopropylidenebis 2,6-dibromophenol, firemaster bp 4a, tetrabromodian, 2,2-bis 3,5-dibromo-4-hydroxyphenyl propane, fire guard 2000, great lakes ba-59p
PubChem CID 6618
ChEBI CHEBI:33217
IUPAC Name 2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC(=C(C(=C1)Br)O)Br)C2=CC(=C(C(=C2)Br)O)Br

Specifications

Melting Point 180°C
Color White
Boiling Point 316°C
Quantity 500 g
UN Number 3077
Formula Weight 543.88
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromobisphenol A
EINECSNumber (4)-0205
RTECSNumber SM0894500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.