Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrabromophenol blue | 4430-25-5 | MFCD00005876 | 98.2% | 985.54 g/mol | C19H6Br8O5S | 250 MG
SDP

Catalog No. 5000283483
Encompass_Preferred

Tetrabromophenol blue (TBPB, IND) is a biochemical indicator dye used in laboratory assays and staining applications. It is supplied as a solid reagent for research use; handle and store according to the safety data sheet and laboratory guidelines.

  • High purity (98.24%) suitable for analytical use.
  • Chemical formula C19H6Br8O5S; molecular weight 985.54 g/mol.
  • Off-white to light yellow solid appearance.
  • Available in small laboratory quantities (250 mg) and larger sizes.
  • Storage: 4°C under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month when stored under nitrogen.
  • Used as a pH indicator and staining reagent in biochemical assays.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-83483 Supplier Medchemexpress LLC Supplier No. HYW140928250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabromophenol blue (TBPB, IND) is a biochemical indicator dye used in laboratory assays and staining applications. It is supplied as a solid reagent for research use; handle and store according to the safety data sheet and laboratory guidelines.

  • High purity (98.24%) suitable for analytical use.
  • Chemical formula C19H6Br8O5S; molecular weight 985.54 g/mol.
  • Off-white to light yellow solid appearance.
  • Available in small laboratory quantities (250 mg) and larger sizes.
  • Storage: 4°C under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month when stored under nitrogen.
  • Used as a pH indicator and staining reagent in biochemical assays.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.