Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrabromophenol blue | 4430-25-5 | MFCD00005876 | 98.2% | 985.54 g/mol | C19H6Br8O5S | 500 MG
SDP

Catalog No. 5000283587
Encompass_Preferred

Tetrabromophenol blue is a biochemical pH indicator dye supplied as a solid for laboratory research use. It is used as a pH indicator and analytical reagent in biochemical assays, and is provided with high purity and specific storage recommendations to maintain stability.

  • Used as a pH indicator in biochemical and analytical assays.
  • High purity (≈98.2%) suitable for research applications.
  • Off-white to light yellow solid form for easy handling.
  • CAS number 4430-25-5 for unambiguous identification.
  • Recommended storage under nitrogen at 4°C; in solvent store at -80°C (long term) or -20°C (short term).

Catalog No. 50-002-83587 Supplier Medchemexpress LLC Supplier No. HYW140928500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabromophenol blue is a biochemical pH indicator dye supplied as a solid for laboratory research use. It is used as a pH indicator and analytical reagent in biochemical assays, and is provided with high purity and specific storage recommendations to maintain stability.

  • Used as a pH indicator in biochemical and analytical assays.
  • High purity (≈98.2%) suitable for research applications.
  • Off-white to light yellow solid form for easy handling.
  • CAS number 4430-25-5 for unambiguous identification.
  • Recommended storage under nitrogen at 4°C; in solvent store at -80°C (long term) or -20°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.