Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenol Blue Disodium Salt, TCI America™
SDP

Catalog No. T00401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T00401G Supplier TCI America Supplier No. T00401G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Formula C19H6Br8O5S
MDL Number MFCD00070515
Synonym Tetrabromophenoltetrabromosulfophthalein Disodium Salt

Specifications

Color Purple/Red
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 1029.51
Chemical Name or Material Tetrabromophenol Blue Disodium Salt
EINECSNumber (5)-3568
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.