Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenol Blue 95.0+%, TCI America™
SDP

Catalog No. T00395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00395G 5 g
T00391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00395G Supplier TCI America Supplier No. T00395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.544
MDL Number MFCD00005876
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym Tetrabromophenoltetrabromosulfophthalein, TBPB
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 204°C
Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 985.54
Percent Purity ≥95.0% (HPLC)
Chemical Name or Material Tetrabromophenol Blue
EINECSNumber (4)-0906&(5)-3568
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.