Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenol Blue, Water Soluble, Thermo Scientific™

Catalog No. AC216780010
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC216780010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC216780010 Supplier Thermo Scientific Chemicals Supplier No. 216780010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
MDL Number MFCD00005876
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Molecular Weight (g/mol) 985.544
PubChem CID 78159
Formula Weight 1007.58
Quantity 1g
Chemical Name or Material Tetrabromophenol Blue
Show More Show Less
EINECSNumber 224-622-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.