Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tetrabromophenolphthalein ethyl ester, pure

Catalog No. AC420070250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420070250 25 g Glass bottle
AC420070010 1 g Glass bottle
AC420070050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC420070250 Supplier Thermo Scientific Chemicals Supplier No. 420070250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1176-74-5 | C22H14Br4O4 | 661.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1176-74-5
Molecular Formula C22H14Br4O4
Molecular Weight (g/mol) 661.96
MDL Number MFCD00066387
InChI Key SQFXATUXPUCFFO-UHFFFAOYSA-N
Synonym tetrabromophenolphthalein ethyl ester, 3',3,5',5-tetrabromophenolphthalein ethyl ester, ethyl tetrabromophenolphthalein, unii-6d91ns781o, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl benzoate, 3 inverted exclamation marka,3 inverted exclamation marka,5 inverted exclamation marka,5 inverted exclamation marka-tetrabromophenolphthalein ethyl ester, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, benzoic acid,2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-,ethyl ester, ethyl 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene methyl benzoate
PubChem CID 92873
IUPAC Name ethyl 2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzoate
SMILES CCOC(=O)C1=CC=CC=C1C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br

Specifications

Melting Point 208.0°C to 211.0°C
Color Red to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9331
Solubility Information Solubility in water: negligible.
Spectrophotometry 80.0% min.
Formula Weight 661.96
Grade Pure
Physical Form Powder
Chemical Name or Material Tetrabromophenolphthalein ethyl ester
Show More Show Less
EINECSNumber 214-645-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.