Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenolphthalein,MP Biomedicals

Catalog No. ICN201437 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201437 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201437 Supplier MP Biomedicals, Inc Supplier No. 0520143710

Chemical Identifiers

CAS 76-62-0
Molecular Formula C20H10Br4O4
Molecular Weight (g/mol) 633.912
InChI Key OBRGVMYQZVQHGO-UHFFFAOYSA-N
PubChem CID 65569
IUPAC Name 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC2(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Formula Weight 633.94
Quantity 10 g
Chemical Name or Material TetrabromoPhenolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.