Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Regis Technologies Inc Tetrabutylammonium Phosphate Buffer, 0.5 M, pH 7.5, 50 mL, clear, colorless liquid
SDP

Catalog No. NC1877911
Encompass

Regis Technologies Inc. Tetrabutylammonium Phosphate Buffer is a high-purity, 0.5 M aqueous solution designed as a complementary ion pair reagent to enhance the resolution of negatively charged analytes in High-Performance Liquid Chromatography (HPLC) applications. This clear, colorless liquid is formulated with a controlled pH of 7.5, ensuring consistent performance in analytical separations.

  • Chemical name: Tetrabutylammonium Phosphate Buffer
  • Concentration: 0.5 M
  • pH: 7.5
  • CAS number: 5574-97-0
  • Molecular formula: C16H38NO4P
  • Molecular weight: 339.46 g/mol
  • Appearance: Clear, colorless liquid
  • Density: 1.008 g/mL
  • Boiling point: Slightly above 100 °C
  • Primary use: Ion pair reagent for HPLC
  • Storage conditions: Tightly closed in a cool, dry, and well-ventilated place
  • Package size: 50 mL

Catalog No. NC1877911 Supplier Regis Technologies Inc Supplier No. 1680505200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Regis Technologies Inc. Tetrabutylammonium Phosphate Buffer is a high-purity, 0.5 M aqueous solution designed as a complementary ion pair reagent to enhance the resolution of negatively charged analytes in High-Performance Liquid Chromatography (HPLC) applications. This clear, colorless liquid is formulated with a controlled pH of 7.5, ensuring consistent performance in analytical separations.

  • Chemical name: Tetrabutylammonium Phosphate Buffer
  • Concentration: 0.5 M
  • pH: 7.5
  • CAS number: 5574-97-0
  • Molecular formula: C16H38NO4P
  • Molecular weight: 339.46 g/mol
  • Appearance: Clear, colorless liquid
  • Density: 1.008 g/mL
  • Boiling point: Slightly above 100 °C
  • Primary use: Ion pair reagent for HPLC
  • Storage conditions: Tightly closed in a cool, dry, and well-ventilated place
  • Package size: 50 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.