Learn More
Medchemexpress LLC Tetrabutylammonium 10g
SDP
Catalog No. 5000206451
Encompass_Preferred
Tetrabutylammonium triiodideIt is an organic compound and belongs to the category of quaternary ammonium salts It is a strong oxidizing agent and iodinating reagent often used in organic chemical reactions especially the conversion of alkenes and alkynes to the corresponding diiodide or tetraiodide compounds Tetrabutylammonium triiodideThere are various applications in synthetic organic chemistry especially in the synthesis of natural products pharmaceuticals and specialty chemicals
Catalog No. 50-002-06451 Supplier Medchemexpress LLC Supplier No. HYW09979210G
Tetrabutylammonium triiodideIt is an organic compound and belongs to the category of quaternary ammonium salts It is a strong oxidizing agent and iodinating reagent often used in organic chemical reactions especially the conversion of alkenes and alkynes to the corresponding diiodide or tetraiodide compounds Tetrabutylammonium triiodideThere are various applications in synthetic organic chemistry especially in the synthesis of natural products pharmaceuticals and specialty chemicals