Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium acetate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012376 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2376 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2376 Supplier MilliporeSigma Supelco Supplier No. 8683525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 301.51
MDL Number MFCD00043208

Specifications

Melting Point 95°C to 98°C
CAS 10534-59-5
Linear Formula (CH3 CH2 CH2 CH2)4 N(OCOCH3)
Quantity 25 g
Solubility Information Acetonitrile: 0.1 g/mL, clear, colorless to very faintly brown
Formula Weight 301.51
Percent Purity ≥99.0% (NT); ≥99.0%
Grade For electrochemical analysis
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.