Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) Complex 98.0+%, TCI America™
SDP

Catalog No. T1272100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1272100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1272100MG Supplier TCI America Supplier No. T1272100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68401-88-7
Molecular Formula C22H36NNiS10
Molecular Weight (g/mol) 693.83
MDL Number MFCD00059124
InChI Key RORVWUFVFQXASJ-UHFFFAOYSA-J
PubChem CID 15397663
IUPAC Name nickel(3+);2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Ni+3]

Specifications

Color Black
Quantity 100 mg
Formula Weight 693.83
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III) Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.