Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Bisulfate Solution

Catalog No. 72453250MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-453-250MLF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 72-453-250MLF Supplier Sigma Aldrich Supplier No. 72453250MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylammonium bisulfate (Bu4NHSO4) can be used as a reagent along with potassium fluoride dihydrate to synthesize +a-fluoro amine derivatives by the ring-opening reaction of aziridines. It may also be utilized as a catalyst in the hydrolysis of aziridines and epoxides to corresponding +a-amino alcohols and 1,2-diols, respectively.

Specifications

CAS 395083-14-4
Melting Point 274°C to 280°C
Molecular Formula C3H5BF3K
Linear Formula C3H5BF3K
MDL Number MFCD11052654
Quantity 5 g
Molecular Weight (g/mol) 147.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.