Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium bisulfate solution, For ion pair chromatography, MilliporeSigma™ Supelco™

Catalog No. 111013924 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3924 6 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3924 Supplier MilliporeSigma Supelco Supplier No. 984796X1AMPF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Concentrate for 6 x 1 L ready-to-use solution. Dilute contents of one ampule (∼0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011637
Synonym Tetrabutylammonium hydrogen sulfate solution

Specifications

CAS 32503-27-8
Packaging Ampule
Linear Formula (CH3 CH2 CH2 CH2)4 N(HSO4)
Quantity 6 x 25 mL
Formula Weight 339.53
Grade For ion pair chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.